Description
- Thick, hard-wearing cotton interlock safety glove
- Palm and fingers with special NBR coating (nitrile rubber)
- Good grip in damp, wet and oily areas
- Good tactile feel
- Ergonomic fit
- High flexibility
- Excellent wearer comfort with the high water vapor absorption of the cotton lining
- Free of harmful substances in line with OEKO-Tex Standard 100
Specifications
Specifications
| Certifications/Compliance | EN 388:2016 (2 1 2 1 X) |
| Coating Material | NBR |
| Color | Orange, White |
| Exterior Finish | Smooth |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Cotton |
| Product Type | Mechanical Protection Gloves |
| Size | 7 |