Learn More
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Product Code. 10126583 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 45 pieces
800 pieces
For 240 samples
Unit Size:
Each
Pack of 800
Pack of 900
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10126583 For 240 samples Each
10715156 800 pieces Pack of 800
10393175 20 x 45 pieces Pack of 900
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10126583 Supplier Thermo Scientific™ Supplier No. 97002141
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
In Stock
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) KingFisher mL Combi 240
Quantity For 240 samples