Learn More
Fisherbrand™ Connecting Cables
Product Code. 11746348 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11746348 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11746348
  • ONLINE EXCLUSIVE PRICE
    45.05 EUR
    40.17 EUR / Pack of 1
    Save 4.88 EUR
    10% Off
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
  • Connecting Cable with BNC Plug
  • For combined ion-sensitive electrode with solid membranes
  • For use with measuring agriculture, water analysis, and calibration of ionic electrodes
Specifications
Concentration 1000 ppm
Connector Types BNC
Type Connecting Cable
For Use With (Application) Agriculture and Water Analysis, Calibration of Ionic Electrodes
Includes Electrolyte Solution Ring
Temperature (Metric) Operating 0°C to 50°C