Learn More
Fisherbrand™ Connecting Cable
Shop All Fisherbrand Products
Click to view available options
Connector Types:
BNC
Banana
DIN
Unit Size:
Each
Specifications
Specifications
| Connector Types | DIN |
| Type | Connecting Cable |
| For Use With (Application) | For connecting the electrode to a pH meter, using an anti -static coaxial cable with the correct plug. |
| Temperature (Metric) Operating | 0°C to 50°C |
| Length (Metric) | 1 m |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur