Learn More
Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug
Product Code. 11866114
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11866114 250 mL Each
11806214 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11866114 Supplier Lenz Laborglasinstrumente™ Supplier No. 04454904
  • ONLINE EXCLUSIVE PRICE
    91.00 EUR
    70.08 EUR / Pack of 1
    Save 20.92 EUR
    22% Off
Estimated Shipment: 04-01-2027
Made of borosilicate glass. Lenz Laborglasinstrumente™ Conical Dropping Funnels with Glass Plug is used to manage the slow transfer of fluids with the featured stopcock to control the flow rate.
  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Pore Size 4.0 mm
Product Type Dropping Funnel