Learn More
Congo Red, MP Biomedicals™
Product Code. 11422092 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422092 5 g 5g
11401852 100 g 100g
11442092 100 g 100g
11411852 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11422092 Supplier MP Biomedicals Supplier No. 0215071105
  • ONLINE EXCLUSIVE PRICE
    28.70 EUR
    27.30 EUR / 5g
    Save 1.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Chemical Identifiers
CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym Cotton Red, Benzo Congo Red
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
Specifications
Formula Weight 696.7
Color Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Congo red