Learn More
Brady™ Cone™ Weather-resistant Blank Cones
Product Code. 11791925 Shop All Brady Products
Change view
Click to view available options
Color:
Orange
Yellow
Height (Metric):
635 mm
889 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Height (Metric) Unit Size
11791925 Yellow 635 mm Each
11711935 Orange 889 mm Each
11721935 Yellow 889 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11791925 Supplier Brady™ Supplier No. 077198
  • ONLINE EXCLUSIVE PRICE
    76.20 EUR
    71.12 EUR / Pack of 1
    Save 5.08 EUR
    6% Off
In Stock
Made of high impact, weather-resistant materials, the Cone™ Warning System is an ideal solution for your indoor or outdoor hazardous warning needs. Cone™ Weather-resistant Blank Cones are four-sided, so your message can't be missed.
  • Made of high impact, weather-resistant materials.
  • Ideal for your indoor or outdoor hazardous warning needs.
  • Cones are four-sided, so your message can't be missed.
Specifications
Alternate Part Number CONE-4SIDE-25H-YELLOW-BLANK
Displayed Text Blank
Height (English) 25 in.
Height (Metric) 635 mm
Length (English) 10.5 in.
Length (Metric) 266.7 mm
Packaging Quantity 1 Ea.
Color Yellow
Material Plastic
Type Blank Cone