missing translation for 'onlineSavingsMsg'
Learn More

Conduritol B Epoxide, 98%, MP Biomedicals™

Product Code. 11474142 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474142 25 mg 25mg
11464142 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474142 Supplier MP Biomedicals Supplier No. 0215889325
  • ONLINE EXCLUSIVE PRICE
    901.00 EUR
    857.85 EUR / 25mg
    Save 43.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6090-95-5
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
InChI Key ZHMWOVGZCINIHW-SCCKRLBHSA-N
Synonym 1r,2r,3s,4r,5r,6s-7-oxabicyclo 4.1.0 heptane-2,3,4,5-tetrol
PubChem CID 25797774
IUPAC Name (1R,2R,3S,4R,5R,6S)-7-oxabicyclo[4.1.0]heptane-2,3,4,5-tetrol
SMILES C1(C(C(C2C(C1O)O2)O)O)O

Specifications

Color White
Density 1.93g/cm3 (L)
Assay Percent Range 0.98
Quantity 25 mg
Formula Weight 162.14
Physical Form Powder
Chemical Name or Material Conduritol B Epoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.