Learn More
Conductivity Solution, 50μS.cm-1, Fisher Chemical™
BIOPHARMA
Product Code. 12954485 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12954485 500 mL 500mL
12944485 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12954485 Supplier Fisher Chemical Supplier No. CTE 004 500
  • ONLINE EXCLUSIVE PRICE
    129.00 EUR
    127.30 EUR / 500mL
    Save 1.70 EUR
    1% Off
 Restricted Item
Estimated Shipment: 29-10-2026
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Conductivity 50μS.cm-1
Physical Form Liquid
Quantity 500 mL