Learn More
Notrax Conductive Runner™ for Rapid Removal of Static Charges
Product Code. 17702258
Change view
Click to view available options
Dimensions (L x W):
1829 x 91 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17702258 1829 x 91 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17702258 Supplier Notrax Supplier No. 743C3660BL
  • ONLINE EXCLUSIVE PRICE
    193.46 EUR
    155.92 EUR / Pack of 1
    Save 37.54 EUR
    19% Off
Estimated Shipment: 05-01-2027
Conductive runner for rapid removal of static charges for lining workbenches, shelves, carts during storage and handling of static sensitive components. Pebble top surface is non-directional and easy to wipe clean with recommended 057 Mat Cleaner.
  • Conductive runner dissipates static from conductive items placed on it
  • Protects sensitive components safe from damaging static shocks
  • Pebble top surface that is abrasion resistant and easy to wipe clean with recommended art
  • 057 Mat Cleaner
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
Specifications
Dimensions (L x W) 1829 x 91 cm
Color Black
Length (English) 60 ft.
Length (Metric) 1829 cm
Material Vinyl
Type Industrial and Commercial Matting
Width (Metric) 91 cm