Learn More
Lenz Laborglasintrumente Condensers with Receiver Acc. To Twisselmann
Product Code. 10643242
Change view
Click to view available options
Standard Taper Joint:
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10643242 NS 34/35 Each
10070251 NS 45/40 Each
10161711 NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10643242 Supplier Lenz Laborglasintrumente Supplier No. 05420105
  • ONLINE EXCLUSIVE PRICE
    652.00 EUR
    499.62 EUR / Pack of 1
    Save 152.38 EUR
    23% Off
Estimated Shipment: 29-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Condensers with receiver
  • Acc. to Twisselmann
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) 70 mL
Material Borosilicate Glass 3.3
Standard Taper Joint NS 34/35