Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Product Code. 10498592
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 24/29
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10498592 NS 24/29 Each
10224631 NS 14/23 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10498592 Supplier Lenz Laborglasintrumente Supplier No. 05210403
  • ONLINE EXCLUSIVE PRICE
    69.00 EUR
    54.94 EUR / Pack of 1
    Save 14.06 EUR
    20% Off
Estimated Shipment: 19-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Condensers acc. to Liebig
  • With olives Liebig condenser
  • Acc. to DIN 12576
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Length (Metric) 400 mm
Standard Taper Joint No NS 24/29
Material Borosilicate Glass (3.3)