Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Product Code. 10428792
Change view
Click to view available options
Length (Metric):
400 mm
500 mm
750 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10428792 500 mm Each
10171901 750 mm Each
10326171 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10428792 Supplier Lenz Laborglasintrumente Supplier No. 05210504
  • ONLINE EXCLUSIVE PRICE
    75.00 EUR
    59.24 EUR / Pack of 1
    Save 15.76 EUR
    21% Off
Estimated Shipment: 19-01-2027
Add to basket
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Liebig condenser acc. to DIN 12576 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Tekniske data
Length (Metric) 500 mm
Standard Taper Joint No NS 29/32
Material Borosilicate Glass (3.3)
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.