Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn
Product Code. 11973421
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 19/26
NS 24/29
Length (Metric):
250 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Unit Size
11973421 NS 19/26 250 mm Each
10401834 NS 24/29 250 mm Each
10612995 NS 24/29 400 mm Each
10090654 NS 14/23 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11973421 Supplier Lenz Laborglasintrumente Supplier No. 05223312
  • ONLINE EXCLUSIVE PRICE
    94.00 EUR
    75.15 EUR / Pack of 1
    Save 18.85 EUR
    20% Off
Estimated Shipment: 28-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Allihn condensers acc. to DIN 12581
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Includes Two threaded plastic olives, GL threads 14
Length (Metric) 250 mm
Standard Taper Joint No NS 19/26