Learn More
Lenz Laborglasinstrumente™ Condenser
Product Code. 10603073
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Length (Metric):
160 mm
250 mm
400 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) unitSize
10603073 NS 14/23 160 mm Each
10592493 NS 19/26 160 mm Each
10077720 NS 29/32 250 mm Each
10030501 NS 24/29 250 mm Each
12643830 NS 19/26 250 mm Each
10060142 NS 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10603073 Supplier Lenz Laborglasinstrumente™ Supplier No. 05210211
  • ONLINE EXCLUSIVE PRICE
    57.00 EUR
    39.53 EUR / Pack of 1
    Save 17.47 EUR
    30% Off
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Condenser for condensing vapors.
  • Chemical resistance
  • High temperature resistance
Specifications
Length (Metric) 160 mm
Quantity 1 Ea.
Standard Taper Joint No NS 14/23
Type Condenser