Learn More
Eppendorf™ Concentrator Plus
BIOPHARMA
Product Code. 11692929 Shop All Eppendorf Products
Change view
Click to view available options
Description:
Basic Concentrator Plus
Concentrator Plus
Concentrator Plus Complete
Concentrator Plus w/ Pump
Electrical Requirements:
230 V, 50/60 Hz (EU)
Includes:
Rotor F-45-48-11
Without Rotor
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Electrical Requirements Includes Unit Size
11692929 Concentrator Plus 230 V, 50/60 Hz (EU) Without Rotor Each
10433995 Basic Concentrator Plus 230 V, 50/60 Hz (EU) Rotor F-45-48-11 Each
11672929 Concentrator Plus Complete 230 V, 50/60 Hz (EU) Rotor F-45-48-11 Each
11338600 Concentrator Plus w/ Pump 230 V, 50/60 Hz (EU) Without Rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11692929 Supplier Eppendorf™ Supplier No. 5305000703
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 11-12-2026
Eppendorf Concentrator plus was designed to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench.
  • Comes in 3 system configurations
  • Extremely quiet operation <50 dB(A)
  • Chemical-resistant, maintenance-free PTFE diaphragm pump eliminates the need for changing pump oil
  • Small footprint saves valuable bench space
  • Wide variety of 15 different rotor options to accommodate tubes from 0.2 mL to 50 mL, microplates and PCR plates
  • Motorless induction drive and chemical-resistant, stainless steel chamber for trouble-free operation
  • Can be connected with most commercially available cold traps for use with vapors
  • Coated lid with superior chemical resistance
  • TRUSTED_SUSTAINABILITY
    Specifications
    Includes Without Rotor
    Pump Type Integrated Diaphragm Vacuum Pump
    Type Concentrators
    Voltage 230 V
    Description Concentrator Plus
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Wattage 350 W
    Dimensions (L x W x H) 58 x 33 x 29 cm (12.6 x 20.9 x 11.7 in.)