Learn More
Eppendorf™ Concentrator Plus
BIOPHARMA
Product Code. 10433995 Shop All Eppendorf Products
Change view
Click to view available options
Description:
Basic Concentrator Plus
Concentrator Plus
Concentrator Plus Complete
Concentrator Plus w/ Pump
Electrical Requirements:
230 V, 50/60 Hz (EU)
Includes:
Rotor F-45-48-11
Without Rotor
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Electrical Requirements Includes Unit Size
10433995 Basic Concentrator Plus 230 V, 50/60 Hz (EU) Rotor F-45-48-11 Each
11692929 Concentrator Plus 230 V, 50/60 Hz (EU) Without Rotor Each
11672929 Concentrator Plus Complete 230 V, 50/60 Hz (EU) Rotor F-45-48-11 Each
11338600 Concentrator Plus w/ Pump 230 V, 50/60 Hz (EU) Without Rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10433995 Supplier Eppendorf™ Supplier No. 5305000100
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
In Stock
Eppendorf Concentrator plus was designed to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench.
  • Comes in 3 system configurations
  • Extremely quiet operation <50 dB(A)
  • Chemical-resistant, maintenance-free PTFE diaphragm pump eliminates the need for changing pump oil
  • Small footprint saves valuable bench space
  • Wide variety of 15 different rotor options to accommodate tubes from 0.2 mL to 50 mL, microplates and PCR plates
  • Motorless induction drive and chemical-resistant, stainless steel chamber for trouble-free operation
  • Can be connected with most commercially available cold traps for use with vapors
  • Coated lid with superior chemical resistance
  • TRUSTED_SUSTAINABILITY
    Specifications
    Frequency 50/60 Hz
    Includes Rotor F-45-48-11
    Pump Type Without Pump
    Type Concentrators
    Voltage 230 V
    Description Basic Concentrator Plus
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Height (Metric) 230 mm
    Speed 1400 rpm
    Wattage 150 W
    Width (Metric) 330 mm
    Dimensions (L x W x H) 37 x 33 x 23 cm (12.6 x 14.5 x 9.1 in.)