missing translation for 'onlineSavingsMsg'
Learn More

Concanavalin A, MP Biomedicals™

Product Code. 11402092 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402092 500 mg 500mg
11492082 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402092 Supplier MP Biomedicals Supplier No. 02150710.5
  • ONLINE EXCLUSIVE PRICE
    430.00 EUR
    409.50 EUR / 500mg
    Save 20.50 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Isolated by affinity chromatography on cross-linked dextran. Concanavalin A, MP Biomedicals is pure Canavalia ensiformis lectin (Con A) from Jackbean. It exists as a dimer below pH 5.0, and at pH >7, it exists as a tetramer.

Lectin are used in a wide variety of applications. Con-A is not a glycoprotein and does not contain cysteine residues. Unlike most other lectins, Con-A is a metalloprotein and requires a transition metal ion, such as manganese, plus calcium ions for binding.

  • Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Con-A binds specifically to mannosyl and glucosyl residues of polysaccharides and glycoproteins.
  • Unmodified hydroxyl groups at the C3, C4 and C6 positions of D-glucopyranosyl or D-mannopyranosyl rings may be essential for binding.
  • Each subunit of Con A contains one calcium ion and one manganese ion. Removal of these cations by dialysis under acidic conditions abolishes the carbohydrate-binding activity.
  • Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer. Above pH 7.0 higher aggregates are formed.
  • Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11028-71-0
Synonym Con A

Specifications

Quantity 500 mg
Solubility Information Soluble in water (10mg/mL, slightly hazy, colorless solution).
Formula Weight Monomeric molecular weight of Con-A is 25,500(Lit.)
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • Concanavalin A
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Concanavalin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.