Learn More
3M™ Comply™ Bowie and Dick Autoclave Test Pack
Product Code. 16854273
Change view
Click to view available options
Quantity:
20 Pk.
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16854273 20 Pk. 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16854273 Supplier 3M™ Supplier No. 7000034520
  • ONLINE EXCLUSIVE PRICE
    294.10 EUR
    232.62 EUR / 20 tests
    Save 61.48 EUR
    20% Off
Estimated Shipment: 09-12-2026
The Bowie and Dick test is carried out daily in an empty chamber and is designed to show the adequacy of steam penetration.
  • Easy to detect failures according to ISO 11140 Part 1 and Part 3
  • Test pack will give you peace of mind and prove your sterilizer is working adequately
  • Saves time - obvious colour change allows easy interpretation, simple and easy to use
  • Accurate, consistent, reliable and reproducible
  • Reduced storage space - compact and disposable
  • Cost saving - paper can be re-used for note-taking
  • Irreversible, for safe storage and traceability
  • One pack of 20 sheets will last you a month - saving time on placing orders
Specifications
Certifications/Compliance EN867-4 and ISO 11140 standards
For Use With (Application) For daily autoclave tests
For Use With (Equipment) Autoclave
Product Type Autoclave Test Pack
Features Unique colour chemistry, disposable
Material Paper
Quantity 20 Pk.
Sterilization Indicator Yes
Sterilization Method Steam