Learn More
BRAND™ Compact Burette, BLAUBRAND™, Class AS, DE-M, with Stopcock
Product Code. 16455979 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16455979 10 mL Each
16465979 25 mL Each
16475979 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16455979 Supplier BRAND™ Supplier No. 13963
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
The New Generation of Burettes from BRAND. With PTFE stopcock, fast to dismantle and easy to clean. Simple repair - all individual components replaceable. Boro 3.3, removable PTFE stopcock. To DIN EN ISO 385. Calibrated to deliver (TD, Ex).
  • With removable PTFE stopcock, fast to dismantle and easy to clean
  • Simple repair - all individual components replaceable
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
All BLAUBRAND™ burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Capacity (Metric) 10 mL
Material Borosilicate Glass (3.3)
Graduations 0.02 mL
Includes Burette tube, stopcock with precision tip, one batch certificate
Stopcock Material PTFE
Type Compact Buret
Class Class AS