Learn More
Macherey-Nagel™ Column Protection System
Product Code. 12738032 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
EC Columns 4/2 with 2mm I.D. or 4/3 with 3, 4 or 4.6mm I.D.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12738032 EC Columns 4/2 with 2mm I.D. or 4/3 with 3, 4 or 4.6mm I.D. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12738032 Supplier Macherey-Nagel™ Supplier No. 718966
  • ONLINE EXCLUSIVE PRICE
    171.00 EUR
    162.75 EUR / Pack of 1
    Save 8.25 EUR
    4% Off
View all promotions
In Stock
Suitable for all analytical HPLC columns with 1/16 inch (1.59mm) fittings. Macherey-Nagel™ Column Protection System is an innovative and universal screw-on guard column holder system for use with EC column cartridges with 4mm column length.
System includes cartridge holder, 2 replacement capillaries (0.12mm ID), 3 ferrules, 2 wrenches and a manual.
  • Cartridges filled with specified NUCLEODUR™, NUCLEOSIL™ and NUCLEOSHELL™ HPLC adsorbents
  • Ideal protection for your analytical main column—significant increase in column lifetime
  • Minimized void volume—suitable also for ultra fast HPLC
  • Special ferrules—pressure stability up to 1300 bar (18 850 psi)
  • Visual contamination check—in-time changing of the guard column
  • Guard column length 4mm, ID 2mm (for main columns with 2mm ID) or ID 3mm (for main columns with 3, 4 and 4.6mm ID)
  • UNIVERSAL RP guard columns suitable for all HPLC columns under RP conditions
Specifications
Length (Metric) 4 mm Column Length
Description Universal screw-on guard column holder system, suitable for analytical HPLC and U-HPLC
Material Stainless Steel
For Use With (Equipment) EC Columns 4/2 with 2mm I.D. or 4/3 with 3, 4 or 4.6mm I.D.
Product Type Column Protection System