Learn More
MP Biomedicals™ Collagenase Type I from Clostridium histolyticum
Shop All MP Biomedicals ProductsDescription
The collagenase product is a mixture of enzymes secreted by C. histolyticum, with different products differentiated by the relative ratios of the 10-18 components found in the secreted enzymes. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular material. The action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.
Unit Definition: One unit will liberate peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Contains clostripain, neutral protease and trypsin activities.
Specifications
Specifications
| Concentration | >125 U/mg |
| Color | Brown |
| For Use With (Application) | Preparation of isolated cell suspensions |
| Content And Storage | Store at 4°C. Desiccate. |
| Quantity | 500 mg |
| Product Type | Collagenase Type I |
| Form | Lyophilized |
Safety and Handling
- Collagenase, Type I
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Collagenase
Your input is important to us. Please complete this form to provide feedback related to the content on this product.