missing translation for 'onlineSavingsMsg'
Learn More

Colistin B Sulfate Salt (~90%), TRC

Product Code. 30041248
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041248 0.5 mg 0.50mg
30040422 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041248 Supplier Toronto Research Chemicals Supplier No. TRCC6415150.5MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1338055-88-1
Molecular Formula C52 H98 N16 O13 . H2 O4 S
Molecular Weight (g/mol) 1253.51
Synonym Colistin B sulfate
IUPAC Name N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,9S,12S,15R,18S,21S)-6,9,18-tris(2-aminoethyl)-3-[(1R)-1-hydroxyethyl]-12,15-bis(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-1-oxobutan-2-yl]-6-methylheptanamide;sulfuric acid
SMILES CC(C)CCCCC(=O)N[C@@H](CCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCN)C(=O)N[C@H]1CCNC(=O)[C@@H](NC(=O)[C@H](CCN)NC(=O)[C@H](CCN)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CCN)NC1=O)[C@@H](C)O.OS(=O)(=O)O

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C52H98N16O13.H2O4S/c1-27(2)11-9-10-12-40(71)59-32(13-19-53)47(76)68-42(31(8)70)52(81)64-35(16-22-56)44(73)63-37-18-24-58-51(80)41(30(7)69)67-48(77)36(17-23-57)61-43(72)33(14-20-54)62-49(78)38(25-28(3)4)66-50(79)39(26-29(5)6)65-45(74)34(15-21-55)60-46(37)75;1-5(2,3)4/h27-39,41-42,69-70H,9-26,53-57H2,1-8H3,(H,58,80)(H,59,71)(H,60,75)(H,61,72)(H,62,78)(H,63,73)(H,64,81)(H,65,74)(H,66,79)(H,67,77)(H,68,76);(H2,1,2,3,4)/t30-,31-,32+,33+,34+,35+,36+,37+,38+,39-,41+,42+;/m1./s1
Formula Weight 1252.7173
Percent Purity >95%
Chemical Name or Material Colistin B sulfate salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.