Learn More
Lenz Laborglasintrumente™ Cold trap with Condensate Drain
Product Code. 10420903
Change view
Click to view available options
Volume (Metric):
200 mL
500 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10420903 200 mL Each
10365831 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10420903 Supplier Lenz Laborglasintrumente™ Supplier No. 05355546
  • ONLINE EXCLUSIVE PRICE
    544.00 EUR
    421.14 EUR / Pack of 1
    Save 122.86 EUR
    22% Off
Estimated Shipment: 08-01-2027
Cold trap for direct filling of cooling agents
  • With valve stopcock (with retaining device)
  • For condensate drain
  • Low cooling agent consumption due to double-walled design
  • Apparatus connection: socket NS 29/32
  • Vacuum pump connection: cone NS 29/32
  • Made of DURAN™ tubing
Specifications
Material DURAN Tubing
For Use With (Equipment) Cooling Agents
Includes Valve stopcock (with retaining device) for condensate drain
Quantity 1
Volume (Metric) 200 mL