missing translation for 'onlineSavingsMsg'
Learn More

Colchicine Methanethiosulfonate, TRC

Product Code. 30355206
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355206 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355206 Supplier Toronto Research Chemicals Supplier No. TRCC6400305MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1217821-52-7
Molecular Formula C22 H25 N O6 . C H4 O2 S2
Quantity 5 mg
Synonym 3-Methylsulfonylsulfanyl-N-[(7S)-1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5H-benzo[d]heptalen-7-yl]propanamide, Colchicine methanethiosulfonate
InChI Formula InChI=1S/C22H25NO6.CH4O2S2/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-18(26-2)17(25)11-15(14)16;1-5(2,3)4/h7,9-11,16H,6,8H2,1-5H3,(H,23,24);1H3,(H,2,3,4)/t16-;/m0./s1
SMILES COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC(=O)C.CS(=O)(=O)S
IUPAC Name sulfanylsulfonylmethane;N-[(7S)-1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
Molecular Weight (g/mol) 511.61
Formula Weight 511.1335
Chemical Name or Material Colchicine Methanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.