missing translation for 'onlineSavingsMsg'
Learn More

Colchiceine, TRC

Product Code. 30138737
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138737 50 mg 50mg
17989809 10 mg 10mg
18070900 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138737 Supplier Toronto Research Chemicals Supplier No. TRCC63850050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-27-0
Molecular Formula C21 H23 N O6
Molecular Weight (g/mol) 385.41
Synonym Colchicine Imp. F (EP), N-[(7S,12aRa)-10-Hydroxy-1,2,3-trimethoxy-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]acetamide, N-[(7S)-5,6,7,9-Tetrahydro-10-hydroxy-1,2,3-trimethoxy-9-oxobenzo[a]heptalen-7-yl]-acetamide, (S)-N-(5,6,7,9-tetrahydro-10-hydroxy-1,2,3-trimethoxy-9-oxobenzo[a]heptalen-7-yl)-acetamide, Colchiceine, Benzo[a]heptalene, acetamide deriv., NSC 33411, O10-Demethylcolchicine
IUPAC Name N-[(7S)-10-hydroxy-1,2,3-trimethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES COc1cc2CC[C@H](NC(=O)C)C3=CC(=O)C(=CC=C3c2c(OC)c1OC)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H23NO6/c1-11(23)22-15-7-5-12-9-18(26-2)20(27-3)21(28-4)19(12)13-6-8-16(24)17(25)10-14(13)15/h6,8-10,15H,5,7H2,1-4H3,(H,22,23)(H,24,25)/t15-/m0/s1
Formula Weight 385.1525
Percent Purity >95%
Chemical Name or Material N-[(7S,12aRa)-10-Hydroxy-1,2,3-trimethoxy-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]acetamide (Colchiceine)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.