Learn More
COL4A3 Antibody (COL4A3/9395R), Novus Biologicals Biologicals™
Product Code. 30448909 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30448909 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30448909 Supplier Novus Biologicals Supplier No. NBP345074
Rabbit Monoclonal Antibody
COL4A3 Monoclonal antibody specifically detects COL4A3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Antigen COL4A3
Applications Immunohistochemistry
Classification Monoclonal
Clone COL4A3/9395R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry : 1-2ug/mL
Formulation 10 mM PBS
Gene Alias ATS2, ATS3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), tumstatin
Host Species Rabbit
Immunogen Recombinant fragment (around aa1400-1670) of human COL4A3 protein (Uniprot: P02462, P08572, P29400, P53420). Exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1285
Target Species Human
Content And Storage Store at -20 to -84°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG κ