missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme Q10, Bovine, MP Biomedicals™

Product Code. 11450972 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450972 25 mg 25mg
11440972 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450972 Supplier MP Biomedicals Supplier No. 0219510825

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 303-98-0
Molecular Formula C59H90O4
Molecular Weight (g/mol) 863.37
MDL Number MFCD00042919
InChI Key ACTIUHUUMQJHFO-UPTCCGCDSA-N
Synonym coenzyme q10, ubidecarenone, ubiquinone-10, ubiquinone 50, coq10, neuquinon, ubiquinone, justquinon, emitolon, heartcin
PubChem CID 5281915
ChEBI CHEBI:46245
IUPAC Name 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.