Learn More
Melford Coelenterazine Native
Product Code. 30018202
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018202 1 mg 1mg
30018203 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018202 Supplier Melford Supplier No. C709000.001
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion.
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion. Used for detecting changes in intracellular Ca2+ in cells that have been transfected with apoaequorin cDNA.
Specifications
CAS 55779-48-1
Molecular Formula C26H21N3O3
Quantity 1 mg