missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Sulfate Heptahydrate, Supelco™

Product Code. 15887120
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15887120 250 g 250g
15877120 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15887120 Supplier Supelco Supplier No. 1025560250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-24-1

Specifications

Melting Point 98°C
Quantity 250 g
MDL Number MFCD00149658
Synonym Cobalt(II) sulfate heptahydrate, Cobalt(II) sulfate heptahydrate
Molecular Weight (g/mol) 281.1
Grade Research
Density 1.95 g/cm3 at 20°C
Chemical Name or Material Cobalt(II) Sulfate Heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.