missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride Anhydrous, Sigma-Aldrich™

Product Code. 15866320
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15866320 100 g 100g
15856320 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15866320 Supplier Sigma Aldrich Supplier No. 8025400100
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-79-9

Specifications

Melting Point 735°C
Quantity 100 g
MDL Number MFCD00010938
Synonym Cobalt(II) chloride anhydrous, Cobalt dichloride, Cobalt(II) chloride anhydrous
Molecular Weight (g/mol) 129.84
Grade Research
Density 3.36 g/cm3 at 25°C
Chemical Name or Material Cobalt(II) Chloride Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.