Learn More
3M™ Coarse Anti-Slip Tape
Product Code. 17297780 Shop All 3M Products
Change view
Click to view available options
Width (Metric):
25 mm
50 mm
100 mm
Unit Size:
12 rolls
24 rolls
6 rolls
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
17297780 100 mm 6 rolls
17277780 25 mm 24 rolls
17287780 50 mm 12 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17297780 Supplier 3M™ Supplier No. 7000145703
  • ONLINE EXCLUSIVE PRICE
    860.00 EUR
    481.42 EUR / 6 rolls
    Save 378.58 EUR
    44% Off
Estimated Shipment: 11-01-2027
Coarse Anti-Slip Tape
  • Anti-slip tape for extreme and heavy traffic areas
  • Extra-large abrasive particles resist clogging in muddy or oily conditions
  • Can be painted without losing friction properties
  • Suitable for use on agriculture and forestry equipment, road building, civil engineering, lorries, tractors, construction vehicles, cranes and building equipment
  • High performance adhesive
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 20 m
Packaging Quantity 1/Cs.
Width (Metric) 100 mm
Color Black