Learn More
3M™ Coarse Anti-Slip Tape
Product Code. 17277780
Change view
Click to view available options
Width (Metric):
25 mm
50 mm
100 mm
Unit Size:
12 rolls
24 rolls
6 rolls
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
17277780 25 mm 24 rolls
17287780 50 mm 12 rolls
17297780 100 mm 6 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17277780 Supplier 3M™ Supplier No. 7000145701
  • ONLINE EXCLUSIVE PRICE
    853.00 EUR
    476.94 EUR / 24 rolls
    Save 376.06 EUR
    44% Off
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
Coarse Anti-Slip Tape
  • Anti-slip tape for extreme and heavy traffic areas
  • Extra-large abrasive particles resist clogging in muddy or oily conditions
  • Can be painted without losing friction properties
  • Suitable for use on agriculture and forestry equipment, road building, civil engineering, lorries, tractors, construction vehicles, cranes and building equipment
  • High performance adhesive
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 20 m
Packaging Quantity 1/Cs.
Width (Metric) 25 mm
Color Black