Learn More
Novus Biologicals™ Coagulation Factor III/Tissue Factor Antibody Pair [HRP]
Product Code. 18645010 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Product Code. 18645010
Brand: Novus Biologicals™ NBP2796505Plates
Polyclonal/Monoclonal Antibody Pair
This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor.
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 23.45 pg/mL
Assay Sensitivity 23.45-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Rat Coagulation Factor III/Tissue Factor.
Target Species Rat
For Use With (Application) Sandwich ELISA
Gene Alias CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor
Gene ID (Entrez) 2152
Gene Symbol F3
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 80 ng of recombinant Rat Coagulation Factor III/Tissue Factor.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Rat Coagulation Factor III/Tissue Factor monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Coagulation Factor III/Tissue Factor polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target Coagulation Factor III/Tissue Factor
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.