missing translation for 'onlineSavingsMsg'
Learn More

Clothianidin, TRC

Product Code. 30138476
Change view
Click to view available options
Quantity:
100 mg
500 mg
10 g
Unit Size:
100mg
10g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138476 10 g 10g
30133643 500 mg 500mg
17932725 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138476 Supplier Toronto Research Chemicals Supplier No. TRCC58850010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210880-92-5
Molecular Formula C6 H8 Cl N5 O2 S
Molecular Weight (g/mol) 249.68
Synonym [C(E)]-N-[(2-Chloro-5-thiazolyl)methyl]-N'-methyl-N' '-nitroguanidine, (E)-1-(2-Chloro-1,3-thiazol-5-yl methyl)-3-methyl-2-nitroguanidine, (E)-1-(2-Chloro-1,3-thiazol-5-ylmethyl)-3-methyl-2-nitroguanidine, (E)-1-(2-Chloro-1,3-thiazole-5-ylmethyl)-3-methyl-2-nitroguanidine, Apacz, Arena, Belay, Celero, Clothianidin, Clutch, Clutch (insecticide), Dantop, Dantotsu, Dantotsu 16WSG, Fullswing, NipsIt, NipsIt Inside, Nipsit Inside Insecticide, Poncho, Poncho 250, Poncho 600, TI 435, TM 44401, Takeloc CLMN 10, Takeloc MC 50 Super, Takeloc MC 50E, Takelock MC 50 super, Titan ST, V 10170
IUPAC Name 1-[(2-chloro-1,3-thiazol-5-yl)methyl]-3-methyl-2-nitroguanidine
SMILES CN\C(=N/[N+](=O)[O-])\NCc1cnc(Cl)s1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H8ClN5O2S/c1-8-6(11-12(13)14)10-3-4-2-9-5(7)15-4/h2H,3H2,1H3,(H2,8,10,11)
Formula Weight 249.0087
Percent Purity >95%
Chemical Name or Material Clothianidin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.