Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Specifications
| Chemical Name or Material | Clothianidin |
| Packaging | Glass Bottle |
| CAS | 210880-92-5 |
| Color | White |
| Melting Point | 176.8°C |
| Synonym | (E)-1-(2-Chloro-5-thiazolylmethyl)-3-methyl-2-nitroguanidine |
| Industry Type | Environmental Analysis |
| Molecular Weight (g/mol) | 249.68 |
| Quantity | 100 mg |
| Grade | Analytical Standard |