missing translation for 'onlineSavingsMsg'
Learn More

Clopyralid, TRC

Product Code. 30011268
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011268 10 g 10g
30004464 100 g 100g
17935085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011268 Supplier Toronto Research Chemicals Supplier No. TRCC58735010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1702-17-6
Formula Weight 190.9541
InChI Formula InChI=1S/C6H3Cl2NO2/c7-3-1-2-4(8)9-5(3)6(10)11/h1-2H,(H,10,11)
IUPAC Name 3,6-dichloropyridine-2-carboxylic acid
Molecular Formula C6 H3 Cl2 N O2
Molecular Weight (g/mol) 192
SMILES OC(=O)c1nc(Cl)ccc1Cl
Synonym Picolinic acid, 3,6-dichloro- (7CI,8CI), 3,6-Dichloro-2-pyridinecarboxylic acid, 3,6-Dichloro-2-picolinic acid, 3,6-Dichloropicolinic acid, Cirtoxin, Clopyralid, Dichloro pyridine acid, Diclopyr, Dowco 290, Faworyt, Faworyt 300SL, IWD 3523, Lontrel, Lontrel 100, Lontrel 3, Lontrel 300, Lontrel L, Lornet, Matrigon, Stinger, Transline, Versatill, XRM 3972
Chemical Name or Material Clopyralid

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.