accessibility menu, dialog, popup

UserName

HOONClCl

CAS RN 1702-17-6

IUPAC Name: 3,6-dichloropyridine-2-carboxylic acid
Synonyms: clopyralid acide dichloro-3,6 picolinique acide 3,6-dichloropicolinique
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: HUBANNPOLNYSAD-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(Cl)=CC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)