missing translation for 'onlineSavingsMsg'
Learn More

Clonidine, TRC

Product Code. 30135838
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135838 1 g 1g
30135295 5 g 5g
30010962 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135838 Supplier Toronto Research Chemicals Supplier No. TRCC5871201G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4205-90-7
Molecular Formula C9 H9 Cl2 N3
Molecular Weight (g/mol) 230.09
Synonym 1H-Imidazol-2-amine, N-(2,6-dichlorophenyl)-4,5-dihydro-, 2-Imidazoline, 2-(2,6-dichloroanilino)- (7CI,8CI), N-(2,6-Dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine, 2-(2,6-Dichloroanilino)-2-imidazoline, 2-(2,6-Dichlorophenylamino)-1,3-diazacyclopentene, 2-(2,6-Dichlorophenylimino)imidazolidine, 734571A, Arkamin, Catapres-TTS, Clonidin, Clonidine, Duraclon, M 5041T, SKF 34427, ST 155BS
IUPAC Name N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine
SMILES Clc1cccc(Cl)c1NC2=NCCN2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H9Cl2N3/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9/h1-3H,4-5H2,(H2,12,13,14)
Formula Weight 229.0174
Percent Purity >95%
Chemical Name or Material Clonidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.