missing translation for 'onlineSavingsMsg'
Learn More

Clomiphene citrate salt, MP Biomedicals™

Product Code. 11481581 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481581 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11481581 Supplier MP Biomedicals Supplier No. 0219017410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 50-41-9
Color White to Yellow
Molecular Formula C32H36ClNO8
Quantity 10 g
Synonym clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
InChI Key PYTMYKVIJXPNBD-BTKVJIOYSA-N
SMILES CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O
IUPAC Name 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 598.089
PubChem CID 6420009
Formula Weight 598.1
Physical Form Crystalline Powder
Chemical Name or Material Clomiphene citrate salt
Show More Show Less
compliance-icons
Product Identifier
  • Clomiphene Citrate Salt
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Clomiphene Citrate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.