Learn More
Clomethiazole Hydrochloride, LoGiCal
Product Code. 30460564
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460564 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30460564 Supplier LoGiCal Supplier No. LGCFOR3353.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Clomethiazole Hydrochloride
CAS 6001-74-7
Analyte or Component Names Clomethiazole Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C6 H8 Cl N S . Cl H
Certifications/Compliance ISO 9001
Synonym Thiazole, 5-(2-chloroethyl)-4-methyl-, hydrochloride (1:1), Thiazole, 5-(2-chloroethyl)-4-methyl-, hydrochloride (6CI,7CI,8CI,9CI), 5-(2-Chloroethyl)-4-methylthiazole hydrochloride, Chlormethiazole hydrochloride, Clomethiazole hydrochloride
InChI Formula InChI=1S/C6H8ClNS.ClH/c1-5-6(2-3-7)9-4-8-5;/h4H,2-3H2,1H3;1H
Solution Type Neat
SMILES Cl.Cc1ncsc1CCCl
IUPAC Name 5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 198.11
Quantity 5 mg
Formula Weight 196.9833 g/mol