missing translation for 'onlineSavingsMsg'
Learn More

Clomazone, TRC

Product Code. 17990685
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990685 1 g 1g
17963095 100 mg 100mg
17980905 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17990685 Supplier Toronto Research Chemicals Supplier No. TRCC5869931G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81777-89-1
Molecular Formula C12 H14 Cl N O2
Molecular Weight (g/mol) 239.7
Synonym 2-[(2-Chlorophenyl)methyl]-4,4-dimethyl-3-isoxazolidinone, Centium 360CS, Cirrus, Cirrus 50WP, Clomazone, Command, Command (pesticide), Command 3ME, Command EC, Dimethazone, F 57020, FMC 57020, Gamit, Isooxadiazon
IUPAC Name 2-[(2-chlorophenyl)methyl]-4,4-dimethyl-1,2-oxazolidin-3-one
SMILES CC1(C)CON(Cc2ccccc2Cl)C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H14ClNO2/c1-12(2)8-16-14(11(12)15)7-9-5-3-4-6-10(9)13/h3-6H,7-8H2,1-2H3
Formula Weight 239.0713
Chemical Name or Material Clomazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.