missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, 98%, MP Biomedicals™

Product Code. 11445951 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445951 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445951 Supplier MP Biomedicals Supplier No. 0219034205

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 637-07-0
Color Colorless
Assay Percent Range 98%
Molecular Formula C12H15ClO3
MDL Number MFCD00000615
Quantity 5 g
Synonym clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
InChI Key KNHUKKLJHYUCFP-UHFFFAOYSA-N
SMILES CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
IUPAC Name ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
Molecular Weight (g/mol) 242.70
PubChem CID 2796
ChEBI CHEBI:3750
Formula Weight 242.7
Physical Form Liquid
Chemical Name or Material Clofibrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.