missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin 2,4-Diphosphate, TRC

Product Code. 17991465
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991465 1 mg 1mg
17984745 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991465 Supplier Toronto Research Chemicals Supplier No. TRCC5799951MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309048-48-3
Molecular Formula C18 H35 Cl N2 O11 P2 S
Molecular Weight (g/mol) 584.94
Synonym L-threo-α-D-galacto-Octopyranoside, methyl 7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, 2,4-bis(dihydrogen phosphate), Methyl 7-Chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propylpyrrolidin-2-yl]carbonyl]amino]-2,4-di-O-phosphono-1-thio-L-threo-α-D-galacto-octopyranoside, Clindamycin 2,4-Bisphosphate, Clindamycin Phosphate Imp. I (EP)
IUPAC Name [(2R,3R,4S,5R,6R)-2-[(1S,2S)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4-hydroxy-6-methylsulfanyl-5-phosphonooxyoxan-3-yl] dihydrogen phosphate
SMILES CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](OP(=O)(O)O)[C@@H](O)[C@H]2OP(=O)(O)O

Specifications

InChI Formula InChI=1S/C18H35ClN2O11P2S/c1-5-6-10-7-11(21(3)8-10)17(23)20-12(9(2)19)14-15(31-33(24,25)26)13(22)16(18(30-14)35-4)32-34(27,28)29/h9-16,18,22H,5-8H2,1-4H3,(H,20,23)(H2,24,25,26)(H2,27,28,29)/t9-,10+,11-,12+,13-,14+,15+,16+,18+/m0/s1
Quantity 1 mg
Formula Weight 584.1125
Shipping Condition Room Temperature
Chemical Name or Material Clindamycin 2,4-bisphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.