Learn More
Clentiazem, TRC
Specifications
Specifications
| Chemical Name or Material | Clentiazem |
| CAS | 96125-53-0 |
| Molecular Formula | C22H25ClN2O4S |
| Quantity | 1 mg |
| Synonym | (+)-cis-2-(4-Methoxyphenyl)-3-acetoxy-5-[2-(N,N-dimethylamino)ethyl]-8-chloro-2,3-dihydro-1,5-benzothiazepin-4(5H)-one, (2S,3S)-3-(Acetyloxy)-8-chloro-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one, (2S-cis)-3-(Acetyloxy)-8-chloro-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one |
| InChI Formula | InChI=1S/C22H25ClN2O4S/c1-14(26)29-20-21(15-5-8-17(28-4)9-6-15)30-19-13-16(23)7-10-18(19)25(22(20)27)12-11-24(2)3/h5-10,13,20-21H,11-12H2,1-4H3/t20-,21+/m1/s1 |
| SMILES | COc1ccc(cc1)[C@@H]2Sc3cc(Cl)ccc3N(CCN(C)C)C(=O)[C@@H]2OC(=O)C |
| IUPAC Name | [(2S,3S)-8-chloro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate |
| Molecular Weight (g/mol) | 448.96 |
| Formula Weight | 448.1224 |