missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ ClearKlens Multi Detergent
Description
- Highly concentrated system based on organic solvents and surfactants used for a wide range of applications
- Efficient use on organic soil like emulsions, strong fatty formulations, coatings, and titanium dioxide based solutions
- Low foam properties
- Shelf life of 2 years
Specifications
Specifications
| Product Type | Detergent |
| Form | Liquid |
| pH | 11 |
| Container Type | Canister |
| Volume (Metric) | 20 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?