Learn More
Diversey ClearKlens Cleansinald SC VH9
Product Code. 17803115
Change view
Click to view available options
Certifications/Compliance:
For DE-Germany, DK-Denmark, FI-Finland, FR-France
Unit Size:
1.25 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Certifications/Compliance unitSize
17803115 For DE-Germany, DK-Denmark, FI-Finland, FR-France 1.25 L
17813115 - 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17803115 Supplier Diversey Supplier No. 101107577
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
Concentrated disinfectant used for the cleaning and disinfection of walls, floors and work surfaces and in the Life Sciences industry.
  • Used for the cleaning and disinfection of walls, floors and work surfaces and in the Life Sciences industry
  • This is a double bagged disinfectant, which has been filtered to 0.2um and sterile filled
  • Has a broad spectrum of activity
  • Can be applied as a spray, wiping or wet sweeping without rinse
  • Small items or dismantled equipment may be immersed in a ClearKlens Cleansinald SC solution
  • Broad spectrum of activity
  • Delivered with conformity certificates
Specifications
Product Type Disinfectant Concentrate
Color-coded No
Form Concentrated Liquid
pH >12
Scent Unscented
Sterility Sterile
Appearance Clear Pale Yellow
Autoclavable Not Autoclavable
Certifications/Compliance For DE-Germany, DK-Denmark, FI-Finland, FR-France
Color Clear
Density Relative density (g/cm3; 20°C): 1.04-1.06
Disposable Single-use
Filtered Non-filtered
Flammable No
Volume (Metric) 50 x 25 mL