Learn More
Vitlab™ Clear SAN Beaker
Product Code. 15782076
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15782076 1000 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15782076 Supplier Vitlab™ Supplier No. 44291
  • ONLINE EXCLUSIVE PRICE
    45.00 EUR
    34.22 EUR / Pack of 6
    Save 10.78 EUR
    23% Off
Estimated Shipment: 26-01-2027
Rated FDA food safe. VITLAB™ Clear SAN Beaker can be used for a wide variety of tasks.
  • Clear
  • Food safe
  • With raised scale
  • Easy-grip handle
TRUSTED_SUSTAINABILITY
Specifications
Material SAN
Capacity (Metric) 1000 mL
Height (Metric) 170 mm
Diameter (Metric) 116 mm