Learn More
Cole-Parmer Cleanroom Male Luer Plugs, Straight Adapter
Product Code. 30053090 Shop All Cole Parmer Products
Change view
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Color Unit Size
30053090 Polypropylene Clear Pack of 10
30054378 Kynar Natural Pack of 10
30054116 Nylon White Pack of 10
30053329 Nylon Natural Pack of 10
30053396 CrystalVu (CVU) Clear Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30053090 Supplier Cole-Parmer Supplier No. 5011036
  • ONLINE EXCLUSIVE PRICE
    10.30 EUR
    8.50 EUR / Pack of 10
    Save 1.80 EUR
    17% Off
Estimated Shipment: 08-12-2026
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Material Polypropylene
Inlet Male Luer, Plug
Color Clear