Learn More
Sampling Systems™ Cleaning Coupons for Plastics
Product Code. 15809551 Shop All Sampling Systems Products
Change view
Click to view available options
Material:
HDPE
Nylon
PTFE
PVC
Polypropylene
Dimensions (L x W):
100 x 100 mm
50 x 50 mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Dimensions (L x W) unitSize
15809551 HDPE 50 x 50 mm Each
15899541 HDPE 100 x 100 mm Each
15829551 Nylon 50 x 50 mm Each
15819551 Nylon 100 x 100 mm Each
15849551 Polypropylene 50 x 50 mm Each
15839551 Polypropylene 100 x 100 mm Each
15869551 PVC 50 x 50 mm Each
15859551 PVC 100 x 100 mm Each
15869561 PTFE 50 x 50 mm Each
15859561 PTFE 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15809551 Supplier Sampling Systems™ Supplier No. 8644H50
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Test and validate your cleaning regime with this range of cleaning validation coupons for various plastics.
  • Tests available for HDPE, nylon, PP, PTFE and PVC
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Product Type Cleaning Coupon
Dimensions (L x W) 50 x 50 mm
For Use With (Application) Testing and validating cleaning regime
Material HDPE